Semivolatile Internal Standard for US EPA Method 525
Supplier: VWR Chemicals BDH®
BDH0730-1ML
BDH0730-1MLEA 82.82 USD
BDH0730-1ML
Semivolatile Internal Standard for US EPA Method 525
Standards Environmental Analysis Standards
Semivolatiles standard.
US EPA Method 525 is an analytical method for the monitoring of semivolatiles in drinking water and raw water sources by GC/MS.