Fortification Solution for US EPA Method 524.3
Supplier: VWR Chemicals BDH®
BDH0724-1ML
BDH0724-1MLEA 41.78 USD
BDH0724-1ML
Fortification Solution for US EPA Method 524.3
Standards Environmental Analysis Standards
Volatiles standard.
US EPA Method 524.3 is a GC/MS method for the analysis of purgeable VOCs in finished drinking water. Analytes added to Methanol 524.3 Revision 4 are supplied in a 24-component mix.