Multi-State Hydrocarbon Standard (High Level)
Supplier: VWR Chemicals BDH®
BDH0719-1ML
BDH0719-1MLEA 172.77 USD
BDH0719-1ML
Multi-State Hydrocarbon Standard (High Level)
Standards Environmental Analysis Standards
Semivolatiles standard.
US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.