Vinyl acetate Organic Standard
Supplier: VWR Chemicals BDH®
BDH0718-1ML
BDH0718-1MLEA 38.9 USD
BDH0718-1ML
Vinyl acetate Organic Standard
Standards Environmental Analysis Standards
Volatiles standard.
US EPA Method 524.2 is a general analytical method for the measurement of purgeable organic compounds in water by capillary column gas chromatography/mass spectrometry.
US EPA Method 624 is a general analytical method for the monitoring of VOCs in municipal and industrial wastewater by GC/MS.
US EPA Method 8260 is a general analytical method for the monitoring of VOCs by GC/MS.