2-Chloroethyl Vinyl Ether Organic Standard
Supplier: VWR Chemicals BDH®
BDH0717-1ML
BDH0717-1MLEA 35.17 USD
BDH0717-1ML
2-Chloroethyl Vinyl Ether Organic Standard
Standards Environmental Analysis Standards
Volatiles Standard.
US EPA Method 524 is a GC/MS method for the analysis of purgeable VOCs in finished drinking water. Analytes added to Methanol 524.3 Revision 4 are supplied in a 24-component mix.
US EPA Method 624 is a general analytical method for the monitoring of VOCs in municipal and industrial wastewater by GC/MS.
US EPA Method 8260 is a general analytical method for the monitoring of VOCs by GC/MS.