Method 525.3 Standard
Supplier: VWR Chemicals BDH®
BDH0708-1ML
BDH0708-1MLEA 121.61 USD
BDH0708-1ML
Method 525.3 Standard
Standards Environmental Analysis Standards
Semivolatiles standard.
US EPA Method 525 is an analytical method for the monitoring of semivolatiles in drinking water and raw water sources by GC/MS.