Volatile Organics Combination Mix for US EPA Methods 502.2, 524, 8021, and 8260
Supplier: VWR Chemicals BDH®
BDH0701-1ML BDH0729-1ML
BDH0729-1MLEA 121.27 USD
BDH0729-1ML BDH0701-1ML
Volatile Organics Combination Mix for US EPA Methods 502.2, 524, 8021, and 8260
Standards Environmental Analysis Standards
Volatiles standard.
US EPA Method 502.2 is a general analytical method for the monitoring of halogenated, purgeable VOCs in drinking water and raw water sources by GC megabore capillary column techniques and Hall and Photoionization Detectors (PID).
US EPA Method 524 is a GC/MS method for the analysis of purgeable VOCs in finished drinking water. Analytes added to Methanol 524.3 Revision 4 are supplied in a 24-component mix.
US EPA Method 8021 is a general analytical method for the monitoring of VOCs by GC megabore capillary column techniques and Hall and Photoionization Detectors (PID).
US EPA Method 8260 is a general analytical method for the monitoring of VOCs by GC/MS.