VWR® Plastic Laminate Worksurface
Supplier: VWR, part of Avantor
AMCLWB3536H AMCLWB3130M AMCLWB3548H AMCLWB3530M AMCLWB3572H AMCLWB3536M AMCLWB3572M AMCLWB3560M AMCLWB3530H AMCLWB3160H AMCLWB3136M AMCLWB3172H AMCLWB3148H AMCLWB3136H AMCLWB3130H AMCLWB3160M AMCLWB3148M AMCLWB3548M AMCLWB3172M AMCLWB3560H
AMCLWB3536HEA 615.46 USD
AMCLWB3536H AMCLWB3130M AMCLWB3548H AMCLWB3530M AMCLWB3572H AMCLWB3536M AMCLWB3572M AMCLWB3560M AMCLWB3530H AMCLWB3160H AMCLWB3136M AMCLWB3172H AMCLWB3148H AMCLWB3136H AMCLWB3130H AMCLWB3160M AMCLWB3148M AMCLWB3548M AMCLWB3172M AMCLWB3560H
VWR® Plastic Laminate Worksurface
Furniture Modular Workstations
Laminate worksurfaces are composed of high-pressure plastic laminate.
  • For High-Column worksurface two or three cut-outs accomodate 76" high columns, depending on bench width
  • High-Column worksurfaces may be used with sitting/bench height carts when height-adjustment is required
Each worksurface has three square edges and one full 180-degree wrapped front edge.