RNase Removing Solution, Nucleaseliminator, VWR®
Supplier: VWR Life Science
E891-50ML-PUMP E891-500ML E891-25PK E891-100ML
97063-078EA 26.14 USD
97063-078 97063-076 97064-070 97063-074
RNase Removing Solution, Nucleaseliminator, VWR®
Cleaning Agents Decontaminants
  • Effectively removes and deactivates nuclease contamination even at high concentrations
  • Safe on all laboratory glass and plastic ware
  • Stable at room temperature for at least two years
DNase and RNase contamination ruins expensive samples and time-consuming protocols in research laboratories every day. NucleasEliminator™ can help protect your investment by removing and deactivating even high concentrations of nuclease contamination from glass or plastic surfaces. Simply wipe, immerse or fill the surface or vessel to be decontaminated with NucleasEliminator™, incubate 5 minutes at room temperature and rinse thoroughly with nuclease-free water. NucleasEliminator™ is safe to use on all surfaces at the laboratory bench ranging from glassware, microcentrifuge tubes, pipette tips and electrophoresis units. NucleasEliminator™ is supplied as a ready-to-use liquid. A 100 ml bottle will treat up to 100 microcentrifuge tubes or 25 agarose electrophoresis casting trays.