VWR® Guard Rail Carts, Stainless Steel
Supplier: VWR, part of Avantor
VWR®
726VW 725VW 526VW 730VW 316VW 729VW
97017-716EA 2880.49 USD
97017-716 97017-714 97017-712 97017-720 97017-722 97017-718
VWR® Guard Rail Carts, Stainless Steel
Carts
Guard rails help prevent items from slipping off shelves, eliminating spills and breakage.
  • Stainless steel fabrication
  • Durable
  • Easy to sanitize
Standard handle and leg bumpers protect cart, walls, and doorways from damage
Note: Capacity is the total cart capacity across all of the shelves with the weight evenly dispersed.
All welded stainless steel construction provides durability and easy sanitation. Equipped with two or three shelves, all-swivel casters, and integrated push handle for easy maneuverability.
Información para pedidos: Ships fully assembled.