VWR® Flow Meters, Acrylic
Supplier: VWR, part of Avantor
VWR®
2510A2A12SVVT-VW 2520A4L54SVVT-VW 2520A4A41SVVT-VW 2520A4L48SVVT-VW 2510A2A00SVVT-VW 2510A2L10SVVT-VW 2510A2A01SVVT-VW 2520A4A44SVVT-VW 2530A4A72SVVT-VW 2510A2A03SVVT-VW 2520A4A33SVVT-VW 2520A4A34SVVT-VW 2530A4A65SVVT-VW 2520A4A42SVVT-VW 2510A2A17SVVT-VW 2520A4A39SVVT-VW 2510A2L11SVVT-VW 2520A4A37SVVT-VW 2520A4A30SVVT-VW 2530A4L64SVVT-VW 2530A4A68SVVT-VW 2520A4A43SVVT-VW 2520A4L52SVVT-VW 2510A2A08SVVT-VW 2510A2A14SVVT-VW 2520A4L38SVVT-VW 2520A4L53SVVT-VW 2520A4A31SVVT-VW 2530A4A67SVVT-VW 2520A4L50SVVT-VW 2510A2A02SVVT-VW 2530A4L69SVVT-VW 2520A4A47SVVT-VW 2520A4A40SVVT-VW 2510A2A29SVVT-VW 2520A4L51SVVT-VW 2510A2A06SVVT-VW 2510A2A07SVVT-VW 2530A4A70SVVT-VW 2520A4A36SVVT-VW 2510A2A15SVVT-VW 2520A4L45SVVT-VW 2520A4L55SVVT-VW 2510A2L19SVVT-VW 2510A2A13SVVT-VW
97004-636EA 150.61 USD
97004-636 97004-792 97004-736 97004-768 97004-600 97004-668 97004-604 97004-748 97004-960 97004-612 97004-708 97004-712 97004-952 97004-740 97004-656 97004-728 97004-672 97004-724 97004-696 97004-972 97004-956 97004-744 97004-784 97004-632 97004-644 97004-756 97004-788 97004-700 97004-948 97004-776 97004-608 97004-980 97004-752 97004-732 97004-660 97004-780 97004-624 97004-628 97004-964 97004-720 97004-648 97004-760 97004-796 97004-676 97004-640
VWR® Flow Meters, Acrylic
Flow Meters
These air and water flowmeters are suitable for use with various applications involving air sampling equipment, aquaculture, desalinization equipment, gas analyzers, medical systems, photoprocessing equipment, and water treatment and distribution systems.
  • Stainless steel fittings, valves, and seals
  • Clear acrylic construction
  • Easy-to-read scales
They feature durable, one-piece, clear acrylic construction; a stable, easy-to-read float; threaded brass inserts for quick installation; and easy-to-read English or metric scales
Flowmeters include stainless steel or PVC FNPT fittings, valve, and seals. Stainless steel fittings include Viton® seals. PVC fittings include Buna-N seals.
Easy disassembly and assembly for maintenance.
Información para pedidos: Flowmeters 97004-956 and -968 require a minimum supply pressure of 10psi when valve is specified. Flowmeter 97004-728 uses a Delrin® float. Flowmeter 97004-956 is not available with valves.