BD™ ESwab Collection Kits, BD Diagnostics
Supplier: BD Medical
BD™
220245 220245 220246 220532 220532
95037-744PK 132.03 USD
95037-744 95062-832 95037-746 CA95037-744 CA95062-832
BD™ ESwab Collection Kits, BD Diagnostics
Swabs Swabs in Tubes
Kit is designed to make specimen collection, transport, and storage easier by sustaining aerobic, anaerobic, and fastitious bacteria in one formulation
The kit includes a blue polypropylene screw-cap tube pre-filled with 1mL of Modified Liquid Amies and packaged with a nylon flexible minitip flocked applicator swab. Tubes protect samples for up to 48 hours, whether stored at room temperature or refrigerated. The sample rapidly elutes from the flocked swab when placed in the transport medium for testing.