VWR® 24-Hour Urine Collection Containers
Supplier: VWR, part of Avantor
VWR®
126920 126911 126919 126910
95037-274CS 213.18 USD
95037-274 95037-276 82028-224 82028-222
VWR® 24-Hour Urine Collection Containers
Sample Containers Urine Containers
Leak-resistant closure system assures specimen integrity during collection and transport.
  • Medical-grade amber HDPE
  • Molded graduations
  • Ergonomic design
  • Leak resistant
  • Trace element controlled
Containers are manufactured from 100% virgin medical grade HDPE. The rigid amber plastic filters UV rays. Molded graduations provide accurate volume determination. Containers come in a horizontal space-saving format or a vertical space-saving format. An ergonomic handle provides convenient and balanced handling when the container is empty or filled. All components are constructed of inert plastics that may be disposed of using an incinerator system. Containers include 70mm closures with gasketted seals or an EZ-UP pour spout for pouring directly into the primary testing vessel. Containers with labels feature instructions and a patient identification label. Volume: 3L (0.79gal.).