BD Falcon™ SWUBE™ Collection and Transport System, BD Diagnostics
Supplier: BD Medical
BD Falcon™ SWUBE™
220210 220690 220090 220690 220090 220210 220710 281130 281130
90001-126CS 391.08 USD
CA-90001-094 90001-126 90001-128 CA-90004-028 CA-90001-034 90001-094 CA-90001-126 90004-028 90001-034
BD Falcon™ SWUBE™ Collection and Transport System, BD Diagnostics
Swabs Swabs in Tubes
Combination swabs and tubes provide a simple and effective method of collection, transporting, protecting, and storing specimens
Sterile, individually wrapped tubes are provided without media, but can be used with a wide array of reagents, media, or solutions. The 14.6–15cm (53/4–6”) long shafts reach touch sampling sites. Available with screw caps or friction caps