BD Falcon™ SWUBE™ Pinworm Paddle Collection Device, BD Diagnostics
Supplier: BD Medical
BD Falcon™ SWUBE™
220120 220120
90001-060CS 991.93 USD
CA-90001-060 90001-060
BD Falcon™ SWUBE™ Pinworm Paddle Collection Device, BD Diagnostics
Swabs Swabs in Tubes
A clear plastic paddle housed inside a 17x100mm plastic tube
Specially designed for the collection and transport of suspected pinworm samples. It may also be used for spore and pollen collection. The paddle features a nontoxic adhesive coating that holds the specimen. The paddle is designed to fit directly onto a microscope stage for easy specimen examination