VWR® C-Leg Bench Frame with Top, Double Bay Uprights and 2 Shelves
Supplier: VWR, part of Avantor
14-C103VWR434 14-C100VWR435 14-C101VWR469 14-C103VWR468 14-C101VWR433 14-C103VWR435 14-C101VWR471 14-C101VWR435 14-C100VWR434 14-C100VWR470 14-C101VWR432 14-C101VWR434 14-C100VWR468 14-C101VWR468 14-C103VWR432 14-C100VWR469 14-C103VWR469 14-C100VWR433 14-C103VWR471 14-C101VWR470 14-C103VWR470 14-C103VWR433 14-C100VWR432 14-C100VWR471
89524-394EA 5319.83 USD
89524-394 89524-258 89524-956 89525-026 89524-320 89524-396 89524-960 89524-324 89524-256 89524-892 89524-318 89524-322 89524-888 89524-954 89524-390 89524-890 89525-028 89524-254 89525-032 89524-958 89525-030 89524-392 89524-252 89524-894
VWR® C-Leg Bench Frame with Top, Double Bay Uprights and 2 Shelves
Furniture Workbenches
VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel.
  • Ergonomic and highly adaptable to the needs of the user
  • Flexible, modular design
  • Suitable for heavy loads up to 1100 lbs.
  • Offering a wide range of accessories to meet any need
There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality. The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. VWR® lab benches are height adjustable in manual, motorized and hand crank versions. All bench frames can be equipped with casters to create a mobile workstation solution.
Información para pedidos: Configurations include frame, top, double bay uprights, and 2 x 12"D Shelves. Frame color is light neutral. Top includes black phenolic or black laminate options.