VWR® C-Leg Bench Frame with Top, Single Bay Uprights, and 2 Shelves
Supplier: VWR, part of Avantor
14-C101VWR463 14-C103VWR429 14-C103VWR427 14-C100VWR431 14-C101VWR466 14-C103VWR462 14-C100VWR467 14-C101VWR426 14-C100VWR464 14-C101VWR427 14-C103VWR465 14-C101VWR429 14-C101VWR467 14-C103VWR467 14-C101VWR430 14-C101VWR428 14-C100VWR462 14-C103VWR431 14-C100VWR426 14-C103VWR466 14-C100VWR428 14-C100VWR430 14-C100VWR429 14-C101VWR462 14-C103VWR464 14-C100VWR427 14-C100VWR466 14-C101VWR464 14-C103VWR428 14-C101VWR431 14-C103VWR426 14-C100VWR463 14-C101VWR465 14-C103VWR430 14-C100VWR465 14-C103VWR463
89525-016EA 6771.95 USD
89525-016 89524-942 89525-014 89524-944 89524-306 89524-308 89524-946 89524-948 89524-310 89524-950 89524-312 89524-378 89524-952 89524-314 89524-250 89524-316 89524-876 89525-024 89524-878 89525-022 89525-020 89524-880 89524-242 89524-384 89524-244 89524-882 89524-386 89524-246 89524-884 89524-388 89524-886 89524-248 89525-018 89524-380 89524-240 89524-382
VWR® C-Leg Bench Frame with Top, Single Bay Uprights, and 2 Shelves
Furniture Workbenches
VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel.
  • Ergonomic and highly adaptable to the needs of the user
  • Flexible, modular design
  • Suitable for heavy loads up to 1100 lbs.
  • Offering a wide range of accessories to meet any need
There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality. The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. VWR® lab benches are height adjustable in manual, motorized and hand crank versions. All bench frames can be equipped with casters to create a mobile workstation solution.
Información para pedidos: Configurations include frame, top, single bay uprights, and 2 x 12"D shelves. Frame color is light neutral. Top includes black phenolic or black laminate options.