VWR® C-Leg Bench Frame with Top, Uprights, and Shelf
Supplier: VWR, part of Avantor
14-C103VWR457 14-C101VWR456 14-C103VWR461 14-C101VWR423 14-C100VWR421 14-C100VWR461 14-C101VWR422 14-C100VWR459 14-C100VWR424 14-C103VWR421 14-C101VWR421 14-C101VWR459 14-C103VWR425 14-C101VWR461 14-C100VWR458 14-C101VWR420 14-C101VWR458 14-C103VWR420 14-C100VWR460 14-C103VWR458 14-C103VWR459 14-C103VWR424 14-C100VWR456 14-C100VWR423 14-C101VWR424 14-C103VWR460 14-C100VWR457 14-C101VWR425 14-C103VWR423 14-C103VWR456 14-C103VWR422 14-C100VWR420 14-C100VWR422 14-C101VWR457 14-C101VWR460 14-C100VWR425
89524-866EA 3366.16 USD
89524-866 89524-304 89524-868 89525-012 89525-010 89524-231 89524-374 89524-296 89524-230 89524-376 89524-298 89524-870 89524-232 89524-872 89525-008 89524-234 89524-930 89524-874 89524-236 89525-006 89524-370 89524-372 89524-294 89524-932 89525-004 89524-238 89524-934 89525-002 89524-936 89524-938 89524-300 89524-366 89524-864 89524-368 89524-940 89524-302
VWR® C-Leg Bench Frame with Top, Uprights, and Shelf
Furniture Workbenches
VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel.
  • Ergonomic and highly adaptable to the needs of the user
  • Flexible, modular design
  • Suitable for heavy loads up to 1100 lbs.
  • Offering a wide range of accessories to meet any need
There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality. The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. VWR® lab benches are height adjustable in manual, motorized and hand crank versions. All bench frames can be equipped with casters to create a mobile workstation solution.
Información para pedidos: Configurations include frame, top, single bay uprights, 1 - 12"D shelf. Frame color is light neutral. Top includes black phenolic or black laminate options.