VWR® 10 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR, part of Avantor
VWR®
89523-484A 82028-428A 82028-426A 82028-432A 89523-482A
89523-484APK 81.81 USD
89523-484A 89523-482A 82028-428A 82028-426A 82028-432A
VWR® 10 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Vials
10-425 autosampler screw thread vials feature a wide opening compatible with 6 mm inserts.
  • Choose from all-steel or improved polymeric-metal version
  • Both compatible with all standard or magnetic Headspace autosamplers
  • Excellent sealing characteristics that withstand high pressures of headspace applications
Spring promotes alignment and ensures insert seals against septa to help guard against evaporation loss.