DuraPorter™ Transport Box, Heathrow Scientific®
Supplier: Heathrow Scientific
HS120052 HS120078 HS120077
89511-782CS 1266.2 USD
89511-782 89511-804 89511-802
DuraPorter™ Transport Box, Heathrow Scientific®
Boxes Transport Boxes
Ideal for transporting biological and clinical samples, as well as instruments and products that need to be kept clean and dry under testing environmental conditions
The clarity of the DuraPorter™ box enables users to visibly check sample integrity, so that it can be opened using the right precautions. All boxes feature leak-proof, polycarbonate construction. Lids open a full 180°.