VWR® Hydrogen Generators (Carrier and Fuel Gas)
Supplier: VWR, part of Avantor
H2PEMPD-1100-100 H2PEMPD-1100-175 H2PEMPD-850-175 H2PEMPD-510-175 H2PEMPD-650-175 H2PEMPD-510-100 H2PEMPD-650-100 H2PEMPD-1300-100 H2PEMPD-850-100 H2PEMPD-1300-175
89497-448EA 23640 USD
89497-448 89497-458 89497-446 89497-456 89497-454 89497-464 89497-452 89497-462 89497-450 89497-460
VWR® Hydrogen Generators (Carrier and Fuel Gas)
Gas Generators Hydrogen Generators
The Carrier Grade Hydrogen Gas Generator is a complete system that includes proton exchange membrane cell, internally controlled electro-chemical cell protection system with pump hydration, simple touchscreen user interface, color changing backlit display system, extra-large internal water reservoir, 0.25" compression output connection port, RS485 serial connection port for optional remote monitoring and control system, and a three-year standard system cell warranty.