VWR® Compound Laboratory Microscopes
Supplier: VWR, part of Avantor
VWR®
1100100501021 1101000300291 1101001400572V 1101001400534V 1101001402164V 1101000300522V 1101000300802V 1101001703491V 1101001400392V 1100100501031 1101001702852V 1101000300811V 1101001701491V 1101000300872V 1101002300631 1101001400533V 1101000300291
89404-468EA 1670.24 USD
89404-468 89496-668 89403-332 89403-330 89403-350 89403-472 89403-368 89403-342 89403-326 89404-470 89403-346 89403-370 89403-348 89403-362 89496-656 89403-328 CA19000-100
VWR® Compound Laboratory Microscopes
Microscopes
VWR® Compound Laboratory Microscopes, equipped with Planachromatic Objectives, are ideal for laboratory, routine, and research work.
  • Equipped with planachromatic objectives
  • Ergonomic, anti-fatigue design
  • Siedentopf-type head with a 30° inclination and 360° rotating capabilities
  • One-year warranty on electrical parts
  • Five-year warranty on mechanical parts and optics
The series offers an ergonomic, anti- fatigue design to enable long hours of comfortable observation.
This VWR® Compound Microscope features a Siedentopf type head with a 30° inclination and 360° rotating capabilities. Standard features include, widefield eyepieces WF10X / 20mm, reversed quintuple nosepiece, plan objectives PL 4X, 10X, 40X S, 100X S-Oil, coaxial coarse and fine focusing adjustment, built in low position coaxial mechanical stage, centerable and focusable 1.25 N.A Abbe condenser, iris diaphragm with filter holder, Koehler illumination halogen 12V / 20W with intensity control, main supply 110V UL approved.
Certificaciones: C-UL, UL Listed. CSA certified.
Información para pedidos: Microscope is supplied with all accessories listed above. The microscope is also supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.