Cleanroom Hood, ESD Grid, White Knight
Supplier: White Knight
89495-932 89495-924 89495-930 89495-928 89495-922 89495-910 89495-918 89495-920 89495-926 89495-914 89495-908 89495-912 89495-934 89495-938 89495-916 89495-936
89495-932EA 34.91 USD
89495-932 89495-924 89495-930 89495-928 89495-922 89495-910 89495-918 89495-920 89495-926 89495-914 89495-908 89495-912 89495-934 89495-938 89495-916 89495-936
Cleanroom Hood, ESD Grid, White Knight
Workwear
These reusable hoods are made with a high-density non-linting material that is rated for ISO Class 3+ cleanrooms requiring electrostatic dissipation (ESD)
Hoods have a tunneled elastic back plus horizontal and vertical adjustment snaps. They feature a split-drape design, with the front panel longer than the back. All seams are laser-cut or pre-serged. Thread is silicon-free.
They include a 1% carbon grid and have excellent durability and longevity. Suitable applications include micro-electronics, semi-conductor manufacturing, and nano research.
Información para pedidos: Custom order product with extended lead time. Contact your VWR Sales Representative for more information.