Cleanroom Coverall, ESD Stripe, White Knight
Supplier: White Knight
89495-800 89495-782 89495-806 89495-786 89495-774 89495-798 89495-794 89495-776 89495-796 89495-792 89495-784 89495-788 89495-804 89495-780 89495-790 89495-772 89495-802 89495-778
89495-800EA 133.32 USD
89495-800 89495-782 89495-806 89495-786 89495-774 89495-798 89495-794 89495-776 89495-796 89495-792 89495-784 89495-788 89495-804 89495-780 89495-790 89495-772 89495-802 89495-778
Cleanroom Coverall, ESD Stripe, White Knight
Workwear
Reusable cleanroom coveralls are made with a high-density polyester calendered fabric that resists fluids and bacterial penetration
Coveralls include knit cuffs with 5% carbon Belltron fiber, a full-length, lint-limiting front zipper with two top snaps at neck, and a mandarin collar. The bottom of the legs include three male snaps and one female snap for adjustment, plus three male snaps on the back of the leg for boot attachment. All seams are laser-cut or pre-serged. Thread is silicon-free.
Available in white and navy.
Designed for cleanroom applications up to and including ISO Class 4, the garments also include a 1% carbon stripe for electrostatic dissipation. Applications include pharmaceutical, bio-tech, and medical device manufacturing.
Información para pedidos: Custom order product with extended lead time. Contact your VWR Sales Representative for more information.