Controlled Environment Frock, ESD Stripe, White Knight
Supplier: White Knight
89495-754 89495-756 89495-758 89495-744 89495-766 89495-764 89495-738 89495-748 89495-768 89495-760 89495-746 89495-752 89495-740 89495-762 89495-770 89495-736 89495-750 89495-742
89495-754EA 68.39 USD
89495-754 89495-756 89495-758 89495-744 89495-766 89495-764 89495-738 89495-748 89495-768 89495-760 89495-746 89495-752 89495-740 89495-762 89495-770 89495-736 89495-750 89495-742
Controlled Environment Frock, ESD Stripe, White Knight
Workwear
Reusable zippered frocks are designed for controlled environments and less critical cleanroom applications
Garments feature a mandarin collar with two snaps at top, a wide-pull zipper, and single snap below at bottom. Assembled with double-needle felled seams for added durability and particle migration prevention. All seams are laser-cut or pre-serged, and thread is silicon-free. Available in white and navy.
Made with lightweight yet durable high-density ESD striped fabric, they resist fluid and bacterial penetration. A 1% carbon stripe provides electrostatic dissipation, and knit cuffs include 5% carbon Belltron fiber. Applications include pharmaceutical, bio-tech, and medical device manufacturing.
Información para pedidos: Custom order product with extended lead time. Contact your VWR Sales Representative for more information.