VWR® Heavy-Duty Utility Wire Carts
Supplier: VWR, part of Avantor
VWR®
89410-090 89410-092 89410-078 89410-080 89410-088 89410-086 89410-092 89410-076 89410-088 89410-086 89410-084 89410-096 89410-094 89410-090 89410-074 89410-078 89410-080 89410-082 89410-094 89410-074 89410-082 89410-076 89410-096 89410-084
89410-078EA 426.4 USD
89410-078 CA89410-076 CA89410-088 CA89410-096 CA89410-074 89410-076 89410-088 CA89410-086 CA89410-094 89410-074 89410-096 89410-086 CA89410-084 CA89410-092 89410-094 CA89410-082 89410-084 CA89410-078 89410-092 CA89410-090 CA89410-080 89410-082 89410-090 89410-080
VWR® Heavy-Duty Utility Wire Carts
Carts
Heavy-duty utility wire carts feature bright chrome-plated wire shelves and corner posts with integrated handles.
  • Shelves Have Truss-Reinforced Edges for Exceptional Durability
  • Shelf Adjustability
  • Carts Can Support up to 544 kg (1200 lb.) When Weight Is Evenly Distributed
Posts also feature groove rings in 2.5 cm (1" ) increments for shelf adjustability.
The carts have an overall height of 99.1 cm (39").
Información para pedidos: This cart is supplied with two or three shelves, two inverted U-frame leg/handle assemblies, and four 12.7 cm (5") swivel casters.