VWR® Stereo Zoom Binocular Microscope
Supplier: VWR, part of Avantor
VWR®
1100200500511
89404-478EA 2438.72 USD
89404-478
VWR® Stereo Zoom Binocular Microscope
Microscopes
The VWR® Stereo Zoom Binocular Microscope is a high-performance unit designed for life science, industrial and pharmaceutical inspection, electronic assembly, and bonding operations.
  • Binocular stereo microscope head
  • Widefield eyepieces
  • Transmitted illumination
The microscope features the Greenough Zoom Optical System, a binocular head with 360° rotation, photo/video: 100% [right eyetube], and widefield eyepieces 10X/F.N. 23. The unit offers a 6.7:1 zoom ratio, WD = 113mm, and a magnification range of 0.75X–5X. Addtional standard features include incident/transmitted large working area stand, 12V/10W intensity-controlled halogen incident, and transmitted illumination. Main power supply is 100V–240V.
Información para pedidos: Microscope is supplied with all accessories listed above. The microscope is supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.