VWR® Clinical Laboratory Trinocular Microscope
Supplier: VWR, part of Avantor
VWR®
1100100501051
89404-474EA 2970.1 USD
89404-474
VWR® Clinical Laboratory Trinocular Microscope
Microscopes
The VWR® Clinical Laboratory Microscope features a trinocular Siedentopf type head with a 30° inclination and 360° rotating capabilities.
  • Ergonomic, anti-fatigue design
  • Widefield eyepieces
Light distribution is 100/0, 20/80
Clinical Laboratory Trinocular Microscope features Planachromatic and Phase Contrast objectives that are ideal for research work. These features, combined with the ergonomic, anti-fatigue design allow for long hours of comfortable observation.
Additional standard features include widefield eyepieces WF 10X/20mm, reversed quintuple nosepiece, planachromatic objectives PL 4X, phase contrast planachromatic objectives PLPh 10X, 40X S, 100X S-Oil, coaxial coarse and fine focusing adjustment, built-in low position coaxial mechanical stage, special five-hole turret type Abbe condenser 1.25 N.A. with choices for phase contrast 10X, 40X, 100X, brightfield and darkfield attaching, centering telescope, and Koehler illumination halogen lamp (12V/20W) with intensity control.
Certificaciones: C-UL, UL Listed. CSA certified.
Información para pedidos: Microscope is supplied with all accessories listed above. The microscope is supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.