VWR® Inverted Fluorescence Microscope
Supplier: VWR, part of Avantor
VWR®
1101000200091V 1101000200301V 1101000300331V 1101000200111V 1100100200551 1101000200152V 1101000200172V 1101000200132V 1101000201381V
89403-662EA 22.05 USD
89403-662 89404-464 89403-642 89403-664 89403-652 89403-564 89403-648 89403-658 89403-656
VWR® Inverted Fluorescence Microscope
Microscopes
The VWR® Inverted Fluorescence Microscope is ideal for applications in biology, cell cytology, genetics, oncology, and immunology, and it is also well suited for research, universities, routine laboratory, and medical demands.
  • Ideal for biology, cell cytology, genetics, oncology, and immunology applications
  • Trinocular Siedentopf tube with 45° inclination
  • One-year warranty on electrical parts
  • Five-year warranty on mechanical parts and optics
The VWR® Inverted Fluorescence Microscope features Siedentopf trinocular tube with a 45° inclination (Light Split = 0/100), Wide Focus PL 10X/22mm high eyepoint eyepiece, side facing quintuple nosepiece, CCIS planachromatic objective PL4X, LWD CCIS planachromatic phase objective Ph10X, LWD PL Ph20X, LWD PL Ph40X, ELWD N.A. 0.30 condenser, centerable phase slider with PH1, PH3, and bright field, phase centering telescope, plain stage equipped with attachable mechanical stage with well plate holders (128X86mm), glass and metal stage inserts, 32mm, 54mm, and 65mm Petri dish holders, eXternally mounted centerable 6V/30W Koehler illumination with intensity control, 45mm blue ground glass and green interference filter, power cord, allen heXagonal key, vinyl dust cover, and universal power supply (100–240V). Complete Epifluorescence attachment includes, Epifluorescence attachment with filter cassette, mercury lamp (100W), lamphouse HBO (100W), collector lens and mercury lamp socket, starter unit HBO (100W), and lamp centering tool. DAPI, FITC, and TRITC filter cubes included.
Certificaciones: C-UL, UL Listed. CSA certified.
Información para pedidos: Microscope is supplied with all accessories listed above. The microscope is also supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.