VWR® Trinocular Inverted Microscope
Supplier: VWR, part of Avantor
VWR®
1101000200262V 1101002400452V 1101002300571V 1101002300262V 1101001400121V 1101000300832V 1101000200272V 1101001700543 1101001700391V 1101001700521V 1101000200471V 1101000200412V 1101001402363 1101001401041V 1101001700461V 1101001401262V 1101000200312V 1101000200292V 1101000200551V 1101001400111V 1101000200011V 1101001400632V 1101000200252V 1101000200282V 1101000200581V 1101001700412 1100100200541
89403-624EA 70.35 USD
89403-624 89403-566 89403-634 89403-632 89403-580 89403-508 89403-620 89403-600 89403-588 89403-592 89403-606 89403-630 89496-650 89403-572 89403-590 89403-574 89403-610 89403-626 89403-612 89403-582 89403-616 89403-576 89403-618 89403-622 89403-602 89403-596 89404-462
VWR® Trinocular Inverted Microscope
Microscopes
The VWR® Inverted Microscope is the ideal device for live cell and advanced applications, as well as routine laboratory, research, and educational needs.
  • Ideal for live cell and advanced applications
  • Trinocular Siedentopf tube with 45° inclination
  • One-year warranty on electrical parts
  • Five-year warranty on mechanical parts and optics
The VWR® Inverted Microscope features a Trinocular Siedentopf trinocular tube with 45° inclination (Light Split = 20/80), Wide Focus PL 10x/22mm high eyepoint eyepiece, side facing quintuple nosepiece, CCIS planachromatic objective PL4X, CCIS planachromatic phase objective Ph10x, LWD PL Ph20x, LWD PL Ph40x, ELWD N.A. 0.30 condenser, centrable phase slider with PH1, PH3, and bright field, phase centering telescope, plain stage with attachable mechanical stage with well plate holders (128x86mm), glass and metal stage inserts,32mm, 54mm, 65mm Petric dish holder, externally mounted centerable 6V/30W Koehler illumination with intensity control, 45mm blue ground glass and green interference filter, power cord, allen hexagonal key, vinyl dust cover, and universal power supply (100–240V).
Certificaciones: C-UL, UL Listed. CSA certified.
Información para pedidos: Microscope is supplied with all accessories listed above. The VWR® Inverted Microscope is supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.