VWR® Stereo Basic Halogen 2x–4x Microscope
Supplier: VWR, part of Avantor
VWR®
1100200800511 1101001400972V 1101001400943V 1101000600651V 1101001400673V 1101006400092V 1101001400902V 1101002400543V 1101001400682V 1101002400402V 1101001400812V 1101001400752V
89404-492EA 642.48 USD
89404-492 89403-384 89403-376 89403-394 89403-386 89403-396 89403-382 89403-400 89403-388 89403-398 89403-380 89403-378
VWR® Stereo Basic Halogen 2x–4x Microscope
Microscopes
The VWR® Stereo Basic Halogen 2X–4X Microscope is designed for use in electronics inspection, education, and life science applications.
  • Binocular stereo microscope head
  • Widefield eyepieces
  • Transmitted illumination
The VWR® Stereo Basic Halogen 2X–4X Microscope features a 45° binocular stereo microscope head, widefield eyepieces 10X/F.N. 20, turret objective magnification of 2X–4X, 12V/10W intensity-controlled halogen incident, and transmitted illumination. Main power supply is 100V–240V.
Información para pedidos: Microscope is supplied with all accessories listed above. The microscope is supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.