BRAND BRANDplates® immunoGrade™ Immunoassay Microplates, Nonsterile, BrandTech®
Supplier: Brand
BRANDplates® BRANDplates® pureGrade™ BRANDplates® immunoGrade™
781742 782301 781720 781723 781721 781724 782306 781731 782300 781722 781741 782305 781740
89093-640PK 1358.46 USD
89093-640 89093-750 89093-630 89400-716 89400-718 89093-628 89093-626 89093-748 89093-624 89400-720 89400-722 89093-752 89093-632
BRAND BRANDplates® immunoGrade™ Immunoassay Microplates, Nonsterile, BrandTech®
Microplates Immunology Microplates and Strips
BRANDplates® immunoGrade™ plates are nonsterile plates treated to provide hydrophilic and hydrophobic regions for "high binding" immobiliazation of proteins
  • "High binding" for both hydrophillic and hydrophobic proteins
  • ASNI-SBS formatted
  • 96-well plates embossed for legibility
All BRANDplates® are manufactured to ANSI/SLAS standards 1 to 4-2004 to ensure compatibility for all processes, especially when automation is involved. The 96-Well plates have a blue embossed alphanumeric code for quick and easy well identification.
Made of virgin polystyrene, these plates are suitable for solid-phase immunoassays, including the majority of standard ELISAs.
Información para pedidos: The 96-Well plates are supplied in packs of 100. The 384-Well plates are supplied in packs of 50. The Strip plates are supplied in packs of 100.