VWR® Plain Apron Tables
Supplier: VWR, part of Avantor
P7206M36N P7606 P7141K30N P7301 P7606K36N P7301M30N P7601 P7202K30N P7201M30N P7202K36N P7146M36N P7606M30N P7141K36N P7601M36N P7206 P7601K36N P7602K30N P7601M30N P7606M36N P7141M36N P7101M30N P7106K36N P7206M30N P7201M36N P7306M30N P7146K36N P7106 P7201K36N P7146M30N P7201 P7602K36N P7101M36N P7101K36N P7106M30N P7101 P7306 P7102K30N P7106M36N P7102K36N P7146K30N P7141M30N P7302K30N P7206K36N
89375-264EA 1880.2 USD
89375-264 89375-242 89375-164 89375-162 89375-382 89375-400 89375-246 89375-168 89375-388 89375-244 89375-266 89375-166 89375-320 89375-408 89375-328 89375-260 89375-380 89375-160 89375-396 89375-154 89375-330 89375-374 89375-252 89375-152 89375-272 89375-158 89375-378 89375-312 89375-332 89375-156 89375-310 89375-276 89375-254 89375-316 89375-238 89375-314 89375-412 89375-318 89375-172 89375-150 89375-270 89375-390 89375-170
VWR® Plain Apron Tables
Furniture Tables
Plain apron tables are available in a variety of sizes, with several different top materials.
  • Available in two heights: 30" or 36"
  • Finished with a chemical resistant, earth-friendly UV finish
  • Legs and aprons are constructed of solid red oak, and are smoothly machined
  • Black rubber leg boots which offer protection against scuffing and chemical spills
  • Equipped with non-skid adjustable glides for precise leveling and for prevention of rocking on uneven floors
Aprons are constructed of solid red oak with heavy-duty 13-gauge steel corner braces. 21/4" square solid oak legs, with 5/16" hanger bolts installed, are attached to apron with nuts and washers supplied.
Load capacity: 600 lbs.
Información para pedidos: Shipped KD with tops attached.