ESK® Sampling Kit, with Tube, Swab and Letheen Broth, Puritan Medical Products
Supplier: Puritan Medical Products
25-83004-PDBLB 25-83004-PDLB 25-83010-PDLB 25-83010-PDBLB
89221-702CS 104.35 USD
89221-702 89221-704 89221-706 89221-708
ESK® Sampling Kit, with Tube, Swab and Letheen Broth, Puritan Medical Products
Swabs Swabs for Sampling
Easy-to-use, reliable test kits are available with Neutralizing Buffer, Buffered Peptone Water, Letheen Broth, or Butterfields Solution, in a 4mL or 10mL fill
Letheen Broth is recommended for the neutralization of quaternary ammonium compounds in disinfectant testing.
Pre-filled with 4mL or 10mL Letheen Broth. Shelf life is 18 months. Made in USA.
They come with a small polyester tip attached to a quick-turn, leak-resistant cap to ensure optimum control.
Información para pedidos: Kits are packaged 50 individually wrapped tubes per case. Bulk packaging includes 6 boxes of 50 individually wrapped tubes per box.