VWR® Laboratory Bottles, High-Density Polyethylene
Supplier: VWR, part of Avantor
VWR®
584220 583170 414004-158 414004-154 414004-155 583150 583120 583130 414004-151 414004-152 584230 583140 583110 414004-159 584250 584200 583180 584210 583100 414004-156 414004-157 584240 414004-153 583190
89221-662CS 598.52 USD
89221-662 89221-580 414004-158 414004-154 414004-155 89221-596 89221-590 89221-592 414004-151 414004-152 89221-664 89221-594 89221-588 414004-159 89221-668 89221-658 89221-582 89221-660 89221-586 414004-156 414004-157 89221-666 414004-153 89221-584
VWR® Laboratory Bottles, High-Density Polyethylene
Bottles Plastic Bottles
Translucent, rigid bottles are for general laboratory use and for storing, shipping, and packing liquids.
  • Durable for long-term use
  • High-density polyethylene construction
  • Translucent siding
Can be stored in freezer to −100 °C (−148 °F). Excellent chemical resistance to most acids, bases, and alcohols. Can be used with most corrosives.
Información de suministro: Polypropylene screw closure included.