VWR® Ergonomic Bubble Floor Mats, Gray
Supplier: VWR International
VWR®
IN0213-S IN0314-S IN0208SILVWR IN0204SILVWR IN0307-S IN0317-S IN0212-S IN0410SILVWR IN0309-S IN0220-S IN0308-S IN0306SILVWR IN0203SILVWR IN0407-S IN0419-S IN0420-S IN0312-S IN0406-S IN0303SILVWR IN0207SILVWR IN0209SILVWR IN0414-S IN0214-S IN0318-S IN0206SILVWR IN0417-S IN0319-S IN0205SILVWR IN0215-S IN0218-S IN0211-S IN0204SILVWR IN0405SILVWR IN0313-S IN0320-S IN0416-S IN0409-S IN0219-S IN0216-S IN0408SILVWR IN0210SILVWR IN0413-S IN0305SILVWR IN0415-S IN0203SILVWR IN0411-S IN0310SILVWR IN0418-S IN0316-S IN0412-S IN0217-S IN0315-S IN0404SILVWR IN0311-S IN0304SILVWR
89185-088EA 1037.91 USD
CA89185-018 CA89185-016 89185-088 76318-924 76318-968 76318-926 76318-928 76318-960 76318-962 76318-964 76318-920 76318-922 76318-966 76318-934 89185-030 76318-936 76318-938 76318-970 76318-972 76318-930 76318-974 76318-976 76318-932 89185-026 89185-028 89185-022 89185-066 89185-100 89185-024 76318-946 76318-948 89185-020 76318-906 76318-908 76318-940 76318-942 76318-944 89185-016 89185-018 89185-056 89185-058 89185-052 76318-912 76318-956 89185-096 76318-914 76318-958 89185-054 76318-916 76318-918 89185-090 76318-950 76318-952 76318-910 76318-954
VWR® Ergonomic Bubble Floor Mats, Gray
Floor Mats Anti-Fatigue Floor Mats
The VWR® Bubble Mat is a true ergonomic mat. The VWR Bubble mat has the comfort of a polyurethane mat and the added benefit of easy clean-ability due to its closed-cell/non-porous top surface.
  • Stimulate the body to increase blood circulation and alertness
  • Molded beveled edges eliminate tripping hazards
  • Silcone free, latex free
  • Antistatic and meets ISO Class 5 (Class 100) compliance
  • Easy-to-clean, closed-cell/nonporous top surface
  • Highly resistant to chemicals and UV light
Safety features include molded beveled edges that will never curl up, eliminating tripping hazards. With the VWR® Bubble mat, you will never have overlapping or worn-out connecting mats, which also eliminates tripping hazards.
Constructed of the highest quality using only virgin raw material, this bubble mat has lifetime ergonomic benefits. The VWR® Bubble mat is silicone & latex-free. Using its unique ergonomic designs, this mat prevents common problems encountered in standing environments, while stimulating the body to increase blood circulation and alertness that will reduce sick leave, workers' compensation and quality issues. The VWR® Bubble mat is a solid mat throughout, which means no hollow bubbles that collapse.
The VWR® Bubble mat can be customized to any size and any shape, always in one piece.
Certificaciones: Meets Cleanroom ISO Class 5 (Class 100) compliance.