Rovers® EndoCervex-Brush®, Therapak®
Supplier: Therapak
36840
89171-026CS 1575.64 USD
89171-026
Rovers® EndoCervex-Brush®, Therapak®
Swabs Swabs for Clinical Use
Rovers® EndoCervex-Brush® is a one piece cytology brush designed to optimize cellular yield in Pap smear, Chlamydia, and Gonorrhea testing
The EndoCervex-Brush® provides 600 separate collection bristles for the simultaneous collection of cellular material. The stiff yet flexible brush head is made of polypropylene and has a protective tip to enhance the overall collection process and to reduce injury and bleeding. The brush head is detachable after collection. This device can be used for the conventional Pap smear or in conjunction with the "monolayer prep" processes. Contra-indications: The Rovers® EndoCervex-Brush® should not be used during pregnancy.