BA 410 Elite Compound Upright Microscope, Motic Instruments
Supplier: Motic
Immobiline™
1100100402981 1101001702941 1101000200122 1100100402971 1101001702881 1101001401801 1101001702901 1101000202171 1101001702871 1101001401791 1101001702871 1101001900631 1101001401791 1101001702891 1101001702891 1101001401801 1101001702901 1101001702881
10767-570EA 3735.9 USD
10767-570 CA89168-454 CA97021-902 10767-568 CA89168-458 CA89168-462 89168-466 10025-414 89168-456 89168-460 CA89168-456 CA19000-900 CA89168-460 89168-464 CA89168-464 89168-462 CA89168-466 89168-458
BA 410 Elite Compound Upright Microscope, Motic Instruments
Microscopes
The BA410 Elite is the flagship of Motic's biological microscope series, and is intended for a wide array of laboratory and research applications
Standard with the BA410 Elite series is Motic’s Enhanced Contrast (EC-H) plan achromatic lenses. These are easily upgraded to Motic’s plan fluar or plan apochromatic lenses for further enhanced image fidelity. A large rackless mechanical stage featuring smooth, ball-bearing drive motion, and 1mm graduated scale allows for precise, repeatable positioning of specimen slides. Robust design and exceptional upgradeability ensure that the BA410 Elite will be a work-horse for years to come.
Modular by design, the BA410 Elite is upgradeable with phase contrast, polarizing, epi-fluorescence, and discussion/teaching devices available to further increase the utility of this microscope. The professional focusing, swing-out Abbe condenser offers focused, consistent lighting across the entire magnification range providing exact, publication quality images when coupled with a Moticam camera.
Información para pedidos: Microscope is supplied with Micrometer reticles, phase contrast accessories, polarizing accessories, Darkfield accessories, and Epi-fluorescence attachments. Contact your VWR representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.