Motic AE2000 Inverted Compound Microscope
Supplier: Motic
1101001703281 1101000201641 1101000201391 1101001703251 1101000201661 1101000201651 1101000201641 1101001703271 1100103800042 1100103800016 1101009700092 1101000201391 1101001703251 1101001703281 1100103800029 1101001703261 1101000201661 1101001703271 1101000200292 1101009700092 1100103800025 1101000201651 1101001703091 1101001703261 1101001703091 1101000201621 1101000200292
89168-416EA 46.2 USD
CA89168-402 CA89168-414 CA89168-420 CA89168-398 CA89168-418 89168-416 89168-414 CA89168-396 89306-272 89168-388 CA89168-412 89168-420 89168-398 89168-402 89306-274 89168-400 89168-418 89168-396 19000-688 89168-412 89168-390 CA89168-416 89168-404 CA89168-400 CA89168-404 CA89168-410 CA19000-688
Motic AE2000 Inverted Compound Microscope
Microscopes
The Motic AE2000 Inverted microscope is the ideal instrument for routine live cell inspection in both educational and high-grade professional applications
Motic’s CCIS plan achromatic lenses are engineered with multi-layer coating for improved light transmission, yielding significantly brighter and sharper images. Controls for focus, light intensity, and the optional mechanical stage are easy to reach, allowing uninterrupted viewing during operation. Combining the AE2000 with a Moticam digital camera provides crisp live cell imaging and transforms the AE2000 into a complete analysis and documentation workstation. Designed for routine lab or clinical work, as well as research requirements in pharmaceutical or university laboratories, the AE2000 is ideal for the routine viewing of cell cultures in microbiology, living organisms, and other tissue culture samples.
The AE2000 offers flexibility in live cell imaging with brightfield and phase contrast. Larger culture flasks (up to 180mm) are easily accommodated by removing the lower portion of the condenser from its dovetail mount. Upgrading to LED lighting is easily achieved through a simple plug-in module for additional lighting options.
Información para pedidos: Microscope is supplied with all accessories listed above. Microscope is customizable with various accessories. For all available options, please contact your local Avantor sales representative. All Motic products come with a one-year electrical and five-year mechanical warranty.