Pipet Aid® XP² Ergonomic Pipetting Device, Drummond Scientific
Supplier: Drummond
Pipet-Aid®
4-000-501
89166-464EA 596.68 USD
89166-464
Pipet Aid® XP² Ergonomic Pipetting Device, Drummond Scientific
Pipets Pipet Controllers
Pipet aid portable pipet controller features an updated pump for improved control, a new contoured handle for increased comfort, and a new charging system that charges during pipet use to minimize downtime and extend operation.
  • Controller fully charges in 2.5 hours
  • Portable pipet controller
  • Removable nosepiece
  • Lightweight controller design minimizes fatigue
  • Quiet pipetting operation
The controller's updates make tasks in the laboratory safer and easier.
It accepts a range of one to 100 ml pipets. The new, more powerful controller pump provides great control and quiet pipetting operation. A removable nosepiece cover enables access to the rubber insert, a filter that locks itself, and a replaceable battery without the need for any tools. The lightweight design minimizes the fatigue associated with extended pipet use.
Certificaciones: UL and C-UL listed.
Información para pedidos: This device includes a removable stand, battery charger/power supply, and four extra self-locking filters. It also includes a one-year manufacturer's warranty on all parts and labor due to manufacturing defects.