VWR® Polypropylene Specimen Containers
Supplier: VWR, part of Avantor
VWR®
0501-3100 0450-1111 0480-1101 0450-1101 0480-1111 0450-1100 0480-1100 0501-3101
89125-844CS 243.96 USD
89125-844 89125-842 89107-308 89125-840 89125-850 89107-310 89107-312 89125-846
VWR® Polypropylene Specimen Containers
Sample Containers General Purpose Containers
Sturdy, shatter-resistant containers are manufactured from transparent polypropylene to allow constant sample viewing.
  • Sturdy, shatter-resistant design
  • Transparent siding
  • Polypropylene construction
  • Available in nonsterile or sterile formats
Containers feature molded graduations in 10 mL and 0.5 oz. increments. Polyethylene snap-on lids are sold separately, except where noted. Lids create a secure seal, and offer space for date, name, doctor, and room number molded into the surface. Components are available in nonsterile or sterile formats.
Información para pedidos: Lids fit both 5 and 8 oz. containers. Bulk items are packaged 20 per bag, 25 bags per case.