VWR® General-Purpose Laboratory Labeling Tape
Supplier: VWR, part of Avantor
VWR®
VWR-T24-10-C VWR-T25-6-C VWR-T45-1-C VWR-T25-16-C VWR-T25-2-C VWR-T14-15-C VWR-T34-6-C VWR-T34-8-C VWR-T14-2-C VWR-T35-2-C VWR-T15-1-C VWR-T35-5-C VWR-T24-11-C VWR-T35-1-C VWR-T24-3-C VWR-T15-4-C VWR-T15-2-C VWR-T34-12-C VWR-T25-12-C VWR-T25-4-C VWR-T34-13-C VWR-T25-1-C VWR-T34-3-C VWR-T35-6-C VWR-T14-6-C VWR-T15-6-C VWR-T34-10-C VWR-T45-4-C VWR-T14-1-C VWR-T35-14-C VWR-T24-17-C VWR-T54-1-C VWR-T24-1-C VWR-T24-9-C VWR-T24-14-C VWR-T25-10-C VWR-T25-11-C VWR-T14-10-C VWR-T24-2-C VWR-T15-7-C VWR-T24-16-C VWR-T35-11-C VWR-T25-5-C VWR-T25-8-C VWR-T24-26-C VWR-T14-13-C VWR-T35-3-C VWR-T44-2-C VWR-T14-26-C VWR-T14-9-C VWR-T34-9-C VWR-T55-1-C VWR-T34-1-C VWR-T34-2-C VWR-T14-3-C VWR-T24-27-C VWR-T54-2-C VWR-T44-6-C VWR-T24-4-C VWR-T54-3-C VWR-T44-1-C VWR-T35-13-C VWR-T34-14-C VWR-T24-15-C VWR-T35-4-C VWR-T14-14-C VWR-T34-17-C VWR-T34-11-C VWR-T14-8-C VWR-T25-7-C VWR-T14-5-C VWR-T24-6-C VWR-T45-2-C VWR-T35-7-C VWR-T24-8-C VWR-T34-16-C VWR-T15-3-C VWR-T25-3-C VWR-T24-7-C VWR-T14-17-C VWR-T14-16-C VWR-T45-5-C VWR-T34-26-C VWR-T24-12-C VWR-T14-27-C VWR-T54-6-C VWR-T24-13-C VWR-T35-16-C VWR-T44-5-C VWR-T25-13-C VWR-T54-4-C VWR-T15-5-C VWR-T24-5-C VWR-T14-12-C VWR-T14-4-C VWR-T54-8-C VWR-T34-7-C VWR-T34-15-C VWR-T14-7-C VWR-T34-5-C VWR-T45-3-C VWR-T14-11-C VWR-T34-27-C VWR-T45-6-C VWR-T34-4-C
89097-968CS 55.07 USD
89097-968 89098-034 89098-118 89098-022 89098-026 89097-910 89098-076 89098-080 89097-928 89098-096 89097-954 89098-102 89097-970 89098-094 89097-998 89097-960 89097-956 89098-044 89098-018 89098-030 89098-046 89098-024 89098-070 89098-104 89097-936 89097-964 89098-040 89098-124 89097-916 89098-090 89097-984 89098-132 89097-986 89098-010 89097-976 89098-014 89098-016 89097-900 89097-996 89097-966 89097-982 89098-086 89098-032 89098-038 89097-988 89097-906 89098-098 89098-112 89097-918 89097-942 89098-082 89098-144 89098-058 89098-068 89097-930 89097-990 89098-134 89098-116 89098-000 89098-136 89098-110 89098-088 89098-048 89097-980 89098-100 89097-908 89098-056 89098-042 89097-940 89098-036 89097-934 89098-004 89098-120 89098-106 89098-008 89098-054 89097-958 89098-028 89098-006 89097-914 89097-912 89098-126 89098-060 89097-972 89097-920 89098-140 89097-974 89098-092 89098-114 89098-020 89098-138 89097-962 89098-002 89097-904 89097-932 89098-142 89098-078 89098-052 89097-938 89098-074 89098-122 89097-902 89098-062 89098-128 89098-072
VWR® General-Purpose Laboratory Labeling Tape
Tapes
VWR® General-Purpose Laboratory Labeling Tape sticks to virtually any surface, can be marked with most writing utensils, and reliably withstands extreme temperature variations.
  • Vinyl-coated, soft satin-finish paper
  • Adhesive sticks to virtually any surface
  • Tape removal leaves no residue
  • Available in two rolls sizes
A wide assortment of vinyl-coated, soft satin-finish paper tape that sticks to virtually any surface. It can be written on with pencil, fiber-tip or ball point pen, and lab markers. This durable, self-adhesive tape is ideal for general laboratory use, and is excellent for applications in refrigeration. It can withstand temperatures from -23 to 121°C (-10 to 250°F) and has an application temperature as low as 0°C (32°F). The adhesive backing will peel off easily and leaves no residue.
Información para pedidos: Available in two roll sizes. Rainbow packs contain blue, green, orange, pink, red, yellow, and white tape.