VWR® Dry Block/Incubator Thermometers
Supplier: VWR, part of Avantor
608102511 60810-1411 60170-6611 60810-1611 60170-6411 608101611 60810-1811 608101411 60810-2511 608101811 608102011 60170-6511 60810-2011
10186-072EA 114.6 USD
CA89095-808 10186-072 89095-804 10186-074 CA89095-802 89095-802 10186-076 89095-808 CA89095-806 89095-806 CA89095-804 CA89095-784 89095-784
VWR® Dry Block/Incubator Thermometers
Thermometers Analog Thermometers
Individually serialized and designed for use in all standard heating blocks and water baths.
  • Contains red organic liquid against white back glass
Thermometers can measure temperature of varying ranges depending on model.