VWR® Verification Thermometers
Supplier: VWR, part of Avantor
VWR®
606000811 60600-1111 606001011 606001311 60600-0611 60600-0811 606000611 60600-0511 606000511 60600-1611 60600-0911 60600-1311 606000911 60170-0611 60170-0711 60600-0711 60600-1011 60600-1511 606001111 606000711
89095-690CS 1441.91 USD
89095-690 75831-366 CA89095-692 CA89095-670 CA89095-680 89095-680 CA89095-690 89095-670 89095-692 10186-012 10186-014 89095-686 89095-676 CA89095-684 89095-684 CA89095-678 75831-368 CA89095-676 CA89095-686 89095-678
VWR® Verification Thermometers
Thermometers Analog Thermometers
To verify sample temperatures in freezers, refrigerators, incubators, or ovens with traceability to NIST.
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • Insert thermometer into the bottle and place near the samples for best results
  • Thermometers contain organic liquid; feature permanently fused markings and lead free glass
  • Bottle contains glass beads or organic liquid
  • Manufactured and tested by our USA quality team; individually serialized
Statement of Accuracy indicates accuracy traceable to NIST for every thermometer.
Información para pedidos: Includes multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.