VWR® TraceClean® 40 ml Vials
Supplier: VWR, part of Avantor
VWR® TraceClean®
QLAV15900012 QLFV15900004 QLAV15900024 QLFV15900010 QLAV15900030 QLAV15900006 QLAV15900018 QLFV15900034 QLAV15900036 QLFV15900016
89093-850CS 220.41 USD
89093-850 89093-870 89093-846 89093-836 89093-866 89093-844 89093-842 89093-832 89093-862 89093-874
VWR® TraceClean® 40 ml Vials
Vials
Type I borosilicate glass vials are for use in volatile organic analyses and are prepared in a solvent-free facility to prevent possible volatile contamination.
  • Uniform 40 ml capacity
  • Clear and amber formats available
  • Custody-sealed packaging to ensure content integrity
Vials are available with polypropylene open-top or solid-top caps.
Processed (PC) and Quality-Assured® (QA) vials follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA vials include a Certificate of Analysis in every case. QA vials also include bar codes with lot number and individual container number for complete traceability. Not Processed (NPC) vials do not receive the specialty cleaning process. All cases are custody-sealed to ensure content integrity.
Solid-top caps feature fluoropolymer resin liners. Open-top caps feature ultrasonically bonded 3.1 mm (¹/₈") fluoropolymer resin/silicone septa.