VWR® TraceClean® 60 ml Vials
Supplier: VWR, part of Avantor
VWR® TraceClean®
QLFV15900044 QLAV15900042 QLAV15900040 QLAV15900044 QLFV15900042
89093-872CS 350.18 USD
89093-872 89093-868 89093-858 89093-864 89093-876
VWR® TraceClean® 60 ml Vials
Vials
Type I clear borosilicate glass vials are designed for use in various analyses and in specimen, extract, and standards storage.
  • Borosilicate glass fabrication
  • Supplied with pen-top caps
  • Prepared to avoid volatile contamination
Vials are prepared in a solvent-free facility to prevent possible volatile contamination
Processed (PC) and Quality-Assured® (QA) vials follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA vials include a Certificate of Analysis in every case. QA vials also include bar codes with lot number and individual container number for complete traceability. Not Processed (NPC) vials do not receive the specialty cleaning process. All cases are custody-sealed to ensure content integrity.
Supplied with pen-top caps that feature ultrasonically bonded 3.1 mm (¹/₈") fluoropolymer resin/silicone septa, which aid in volatile organic analyses.