BRAND BRANDplates® cellGrade™ Microplates for Cell Culture, Sterile, BrandTech®
Supplier: BrandTech
BRANDplates®
781982 781981 781962 781960 781961 781965 781971 781968 781986 781970 781980 781987
89093-684PK 342.21 USD
89093-684 89093-680 89093-790 89093-780 89093-690 89093-778 89093-688 89093-776 89093-678 89093-788 89093-686 89093-676
BRAND BRANDplates® cellGrade™ Microplates for Cell Culture, Sterile, BrandTech®
Microplates Cell Culture Plates
BRANDplates® cellGrade™ plates are sterile plates treated for the cultivation of adherent cell lines
  • Cell culture treated surface
  • ANSI-SBS formatted
  • Sterility assurance level of 10-6
All BRANDplates® are manufactured to ANSI/SLAS standards 1 to 4-2004 to ensure compatibility for all processes, especially when automation is involved. The 96-Well plates have an orange embossed alphanumeric code for easy well identification.
The hydrophilic surface has freely accessible carboxyl and hydroxyl groups for the binding of serum components for indirect cell adhesion. All plates are made of virgin polystyrene. Sterilized by beta irradiation according to Ph. Eur. and USP 29, to a Sterility Assurace Level of 10-6.
Información para pedidos: All plates are individually wrapped with lid in packs of 50.