BRAND BRANDplates® pureGrade™ S Microplates, Sterile, BrandTech®
Supplier: BrandTech
BRANDplates®
781665 781661 781668 781660 781663 781670 781662 781686 781681 781680 781687 781682 781671
89093-608PK 305.85 USD
89093-608 89093-618 89093-616 89093-734 89093-614 89093-732 89093-622 89093-612 89093-744 89093-620 89093-730 89093-610 89093-742
BRAND BRANDplates® pureGrade™ S Microplates, Sterile, BrandTech®
Microplates HTS Microplates
BRANDplates® pureGrade™ S plates are sterile, untreated plates designed for a variety of standardized applications
  • Untreated, "medium binding" plates
  • ANSI-SLAS formatted
  • Sterility assurance level of 10-6
All BRANDplates® are manufactured to ANSI/SLAS standards 1 to 4-2004 to ensure compatibility for all processes, especially when automation is involved. White and black 96-Well plates have a gray embossed alphanumeric code for quick and easy well identification.
Made of virgin polystyrene, these plates are suitable for homogenous assays, screening, and storage. Plates are sterilized by beta irradiation according to Ph. Eur. and USP 29, to a Sterility Assurace Level of 10-6.
Información para pedidos: All plates are supplied wth lids in packs of 50.