BRAND BRANDplates® lipoGrade™ Immunoassay Microplates, Nonsterile
Supplier: Brand
BRANDplates®
781840 781862 781849 781851 781842
89093-674PK 1358.48 USD
89093-674 89093-660 89093-670 89093-656 89093-764
BRAND BRANDplates® lipoGrade™ Immunoassay Microplates, Nonsterile
Microplates Immunology Microplates and Strips
BRANDplates® lipoGrade™ plates are nonsterile plates treated for immobilization of predominantly hydrophobic proteins for solid phase assays
  • Binds hydrophobic proteins
  • Maintains hydrophilic proteins in solution
  • ANSI-SBS formatted
All BRANDplates® are manufactured to ANSI/SLAS standards 1 to 4-2004 to ensure compatibility for all processes, especially when automation is involved. The 96-Well plates have a blue embossed alphanumeric code for quick and easy well identification.
Convesely, they are also excellently suited for homogeneous assays of hydrophilic proteins in solution. All plates are made of virgin polystyrene.
Información para pedidos: The 96-Well plates are supplied in packs of 100. The 384-Well plates are supplied in packs of 50.