EZ Grip Tissue Culture Nosepiece, Drummond
Supplier: Drummond
4-000-079-W 4-000-087
89082-452EA 114.89 USD
89082-452 80078-970
EZ Grip Tissue Culture Nosepiece, Drummond
Pipets Pipet Controllers
Drummond EZ grip tissue culture nosepiece eliminates the possibility of cross contamination due to over pipetting.
  • Self-locking action of hydrophilic layer prevents further pipetting with contaminated filter
  • Double-membrane filter
It incorporates a dual-hydrophilic 0.8 μm, hydrophobic 0.8 μm filter, which prevents further use once contaminated by fluid.
Each kit contains all necessary parts including four filters.
This double membrane filter provides a barrier to particles as small as 0.1 μm.
Información accesorios: Extra filters may be ordered separately.
Información para pedidos: The EZ Grip Tissue Culture Nosepiece is for use on Pipet-Aid XP, XL, and Hoodmate only.